Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205486.1 | NP_001192415.1 | 1 - 2 | 341 | 88 | |
GenBank mRNA | BC142221.1 | - | 1 - 2 | 324 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |