Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205543.1 | NP_001192472.1 | 1 - 2 | 62 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |