Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205585.1 | NP_001192514.1 | 4 - 5 | 841 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |