Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205748.2 | NP_001192677.1 | 7 - 8 | 3547 | 95 | |
GenBank mRNA | BC149148.1 | - | 3 - 4 | 1715 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |