Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205815.2 | NP_001192744.2 | 7 - 8 | 905 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |