Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205849.3 | NP_001192778.3 | 31 - 32 | 3947 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |