Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205849.3 | NP_001192778.3 | 4 - 5 | 587 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |