Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205868.1 | NP_001192797.1 | 2 - 3 | 503 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |