Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206026.1 | NP_001192955.1 | 1 - 2 | 116 | 80 | |
GenBank mRNA | AY143368.1 | - | 1 - 2 | 78 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |