Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206054.2 | NP_001192983.1 | 2 - 3 | 735 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |