Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206092.3 | NP_001193021.1 | 4 - 5 | 614 | 125 | |
GenBank mRNA | BC142491.1 | - | 4 - 5 | 602 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |