Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206203.1 | NP_001193132.1 | 2 - 3 | 143 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |