Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192477.1 | NP_001179406.1 | 11 - 12 | 1854 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |