Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192515.2 | NP_001179444.2 | 6 - 7 | 956 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Bos taurus Probe spans exons |