Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192534.1 | NP_001179463.1 | 3 - 4 | 556 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |