Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206279.1 | NP_001193208.1 | 1 - 2 | 45 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |