Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206405.1 | NP_001193334.1 | 6 - 7 | 704 | 81 | |
GenBank mRNA | BC103457.1 | - | 6 - 7 | 694 | 81 | |
BC149976.1 | - | 6 - 7 | 705 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |