Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206474.1 | NP_001193403.1 | 3 - 4 | 339 | 74 | |
GenBank mRNA | BC103434.1 | - | 3 - 4 | 341 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |