Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192677.1 | NP_001179606.1 | 3 - 4 | 645 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Bos taurus Probe spans exons |