Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192677.1 | NP_001179606.1 | 4 - 5 | 769 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Bos taurus Probe spans exons |