Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192820.3 | NP_001179749.1 | 8 - 9 | 1109 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |