Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192875.3 | NP_001179804.1 | 4 - 5 | 638 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |