Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206678.1 | NP_001193607.1 | 5 - 6 | 636 | 62 | |
GenBank mRNA | AY183660.1 | - | 2 - 3 | 169 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |