Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206720.2 | NP_001193649.1 | 2 - 3 | 464 | 69 | |
GenBank mRNA | DV874786.1 | - | 2 - 3 | 464 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |