Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166568.1 | NP_001160040.1 | 2 - 3 | 548 | 108 | |
GenBank mRNA | BC151740.1 | - | 2 - 3 | 403 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |