Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167932.2 | NP_001161404.2 | 1 - 2 | 46 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |