Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167932.2 | NP_001161404.2 | 8 - 9 | 1027 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |