Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172576.2 | NP_001166047.1 | 3 - 4 | 563 | 99 | |
NM_001301322.1 | NP_001288251.1 | 3 - 4 | 560 | 99 | ||
GenBank mRNA | BC149484.1 | - | 3 - 4 | 526 | 99 | |
CO887362.1 | - | 3 - 4 | 520 | 99 | ||
EE350146.1 | - | 3 - 4 | 563 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |