Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172576.2 | NP_001166047.1 | 4 - 5 | 649 | 76 | |
NM_001301322.1 | NP_001288251.1 | 4 - 5 | 646 | 76 | ||
GenBank mRNA | BC149484.1 | - | 4 - 5 | 612 | 76 | |
BC149845.1 | - | 2 - 3 | 115 | 76 | ||
CO887362.1 | - | 4 - 5 | 606 | 76 | ||
EE350146.1 | - | 4 - 5 | 649 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |