Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191217.4 | NP_001178146.2 | 3 - 4 | 362 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |