Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191290.3 | NP_001178219.2 | 7 - 8 | 3919 | 108 | |
GenBank mRNA | EV738882.1 | - | 2 - 3 | 525 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |