Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206822.1 | NP_001193751.1 | 12 - 13 | 1865 | 73 | |
GenBank mRNA | DV877502.1 | - | 3 - 4 | 398 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |