Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206892.2 | NP_001193821.2 | 2 - 3 | 131 | 97 | |
GenBank mRNA | BC151809.1 | - | 2 - 3 | 132 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |