Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206963.1 | NP_001193892.1 | 2 - 3 | 167 | 102 | |
GenBank mRNA | EV628200.1 | - | 2 - 3 | 167 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Probe spans exons |