Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191358.1 | NP_001178287.1 | 3 - 4 | 689 | 88 | |
GenBank mRNA | HQ728434.1 | - | 4 - 5 | 573 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |