Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191443.1 | NP_001178372.1 | 5 - 6 | 1274 | 65 | |
GenBank mRNA | EE249246.1 | - | 3 - 4 | 337 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |