Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191476.2 | NP_001178405.1 | 5 - 6 | 1329 | 81 | |
GenBank mRNA | EV635295.1 | - | 4 - 5 | 487 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |