Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173957.2 | NP_776382.1 | 1 - 2 | 82 | 116 | |
GenBank mRNA | AB035446.1 | - | 1 - 2 | 82 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |