Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192080.2 | NP_001179009.2 | 1 - 2 | 67 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |