Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192105.1 | NP_001179034.1 | 5 - 6 | 770 | 93 | |
NM_001205290.1 | NP_001192219.1 | 6 - 7 | 864 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |