Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192310.2 | NP_001179239.2 | 9 - 10 | 1122 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |