Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192359.1 | NP_001179288.1 | 12 - 13 | 1501 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |