Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192403.1 | NP_001179332.1 | 6 - 7 | 1021 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |