Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192431.1 | NP_001179360.1 | 4 - 5 | 609 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |