Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192912.1 | NP_001179841.1 | 2 - 3 | 514 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |