Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192932.2 | NP_001179861.2 | 9 - 10 | 1027 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |