Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192999.1 | NP_001179928.1 | 17 - 18 | 2272 | 100 | |
GenBank mRNA | BC105208.1 | - | 16 - 17 | 2029 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |