Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193048.2 | NP_001179977.1 | 1 - 2 | 90 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |