Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193201.1 | NP_001180130.1 | 2 - 3 | 240 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |