Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193224.2 | NP_001180153.2 | 1 - 2 | 65 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |